Learn More
Ethanolamine, 98+%
Description
Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.
Solubility
Miscible with water.
Notes
Store it away from heat, spark, open flames, hot surfaces and smoke. The storage area should be safe and enclosed.

Specifications
Specifications
Melting Point | 10°C |
Density | 1.012 |
Boiling Point | 169°C to 170°C |
Flash Point | 93°C (199°F) |
Odor | Ammonia-like |
Refractive Index | 1.454 |
Linear Formula | HOCH2CH2NH2 |
Quantity | 500 g |
UN Number | UN2491 |
Beilstein | 505944 |
Show More |
Safety and Handling
Hazard Category | H227-H302+H312+H332-H314-H335 |
Hazard Statement | GHS H Statement H314-H302-H312-H332-H227 Causes severe skin burns and eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Combustible liquid. |
Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ETHANOLAMINE |
EINECSNumber | 205-483-3 |
RTECSNumber | KJ5775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.