Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanolamine, 99%

Catalog No. AC149580200
Click to view available options
Quantity:
250 mL
1 L
2.5 L
20 L
25 L

CAS: 141-43-5 | C2H7NO | 61.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
SMILES NCCO

Specifications

Density 1.0120g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)
Linear Formula H2NCH2CH2OH
Quantity 20 L
Beilstein 04, 274
Fieser 01,357; 02,189; 04,222; 16,162
Merck Index 15, 3782
IUPAC Name 2-aminoethan-1-ol
Vapor Pressure 0.48mmHg at 20°C
Formula Weight 61.08
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Ethanolamine, 99%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-483-3
RTECSNumber KJ5775000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.