Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanolamine, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
20L
25L
141-43-5
C2H7NO
61.08
MFCD00008183
HZAXFHJVJLSVMW-UHFFFAOYSA-N
ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine
700
CHEBI:16000
2-aminoethanol
C(CO)N
141-43-5
C2H7NO
61.08
MFCD00008183
HZAXFHJVJLSVMW-UHFFFAOYSA-N
ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine
700
CHEBI:16000
2-aminoethanol
C(CO)N
Viscosity 24 mPa.s (20°C)
Infrared Spectrum Authentic
Flash Point 92°C
Density 1.0120g/mL
Boiling Point 170.0°C
Refractive Index 1.4530 to 1.4550
Melting Point 10.3°C
Quantity 250mL
Packaging Glass bottle
Vapor Pressure 0.48mmHg at 20°C
Water 0.5% max.
Color Undesignated
Specific Gravity 1.012
Assay Percent Range 98.5% min. (GC)
Linear Formula H2NCH2CH2OH
Beilstein 04, 274
Fieser 01,357; 02,189; 04,222; 16,162
Merck Index 15, 3782
Solubility Information Solubility in water: soluble. Other solubilities: miscible with methanol and acetone, soluble in benzene, ether, ccl4 and n-heptane
Formula Weight 61.08
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Ethanolamine, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:

GHS Signal Word: Danger

EINECSNumber : 205-483-3

RTECSNumber : KJ5775000

TSCA : TSCA