Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanolamine, 99%, H2O 0.5% max, Thermo Scientific Chemicals

Catalog No. p-7049788
Click to view available options
Quantity:
250 g

Ethanolamine is used as a surfactant, fluorimetric reagent, and to remove CO2 and H2S from natural gas and other gases, functional fluids, intermediates. It is also used as feedstock in the production of detergents, emulsifiers, polishes, pharmaceuticals, corrosion inhibitors, chemical intermediates, ion exchange agents, plating agents and surface treating agents, electrical and electronic products, paper products, personal care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
IUPAC Name 2-aminoethanol
SMILES NCCO

Specifications

Melting Point 10°C
Density 1.012 g/mL
Boiling Point 169°C to 170°C
Flash Point 93°C (199°F)
Odor Ammonia-like
Refractive Index 1.454
Quantity 250 g
UN Number UN2491
Beilstein 505944
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,3727
Solubility Information Soluble in benzene,ether,carbon tetrachloride.
Formula Weight 61.08
Percent Purity 99%
Concentration or Composition (by Analyte or Components) H2O 0.5% max.
Physical Form Liquid
Chemical Name or Material Ethanolamine
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ETHANOLAMINE
EINECSNumber 205-483-3
RTECSNumber KJ5775000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.