Learn More
Ethanolamine, ACS, 99+%, Thermo Scientific Chemicals
Description
It is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 10°C |
Density | 1.012 g/mL |
Boiling Point | 169°C to 170°C |
Flash Point | 93°C (199°F) |
Assay Percent Range | >99+% |
Odor | Ammonia-like |
Refractive Index | 1.454 |
Quantity | 4 x 500 mL |
UN Number | UN2491 |
Beilstein | 505944 |
Show More |
Safety and Handling
Hazard Category | H227-H302+H312+H332-H314-H335 |
Hazard Statement | GHS H Statement H314-H318-H302-H312-H332-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Combustible liquid. |
Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ETHANOLAMINE |
EINECSNumber | 205-483-3 |
RTECSNumber | KJ5775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.