Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanolamine, (CO2 Absorber), Scintillation Grade 99%, Thermo Scientific™

Catalog No. ac409535000
Click to view available options
Quantity:
500 mL

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym 2-Aminoethanol
PubChem CID 700
ChEBI CHEBI:16000
IUPAC Name 2-aminoethan-1-ol
SMILES NCCO

Specifications

Physical Form Liquid
Density 1.01
Quantity 500 mL
Linear Formula NH2CH2CH2OH
Beilstein 04, 274
Fieser 01,357; 02,189; 04,222; 16,162
Merck Index 15, 3782
Formula Weight 61.08
Percent Purity ≥98.5%
Chemical Name or Material Ethanolamine, 99%
Hazard Category Danger
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.