Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETHE1 Antibody (001), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290300PE
Description
ETHE1 Monoclonal antibody specifically detects ETHE1 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
ETHE1 | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
001 | |
Western Blot, ELISA, Immunoprecipitation | |
D83198, EC 3.1.2.6, ethylmalonic encephalopathy 1, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, HSCOEC 3.-, protein ETHE1, mitochondrial, YF13H12 | |
Rabbit immunized with purified, recombinant Human ETHE1 (Uniprot#: O95571, Leu13-Ala254) | |
0.1 mL | |
metabolism | |
23474 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction