Learn More
ETHE1 Antibody (1621), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP243632
Catalog No. NBP243632
Only null left
Add to Cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence
ETHE1 Monoclonal specifically detects ETHE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ETHE1
Monoclonal
Unconjugated
PBS with No Preservative
ETHE1
Full length human ETHE1 Recombinant protein.
0.1 mL
metabolism
23474
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG2b
Western Blot, Immunocytochemistry, Immunofluorescence
1621
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
D83198, EC 3.1.2.6, ethylmalonic encephalopathy 1, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, HSCOEC 3.-, protein ETHE1, mitochondrial, YF13H12
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only