Learn More
Invitrogen™ ETHE1 Monoclonal Antibody (GT1621)

Description
Positive Control: Neuro2A, GL261, C8D30, NIH-3T3, Raw264.7, C2C1, PC-12, Rat2 Predicted Reactivity: Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ETHE1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT1621 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Ethe1 |
| Gene Accession No. | O95571, Q9DCM0 |
| Gene Alias | 0610025L15Rik; ETHE1; ETHE1, persulfide dioxygenase; ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; hepatoma subtracted clone one protein; HSCO; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; sulfur dioxygenase ETHE1; YF13H12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.