Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETHE1 Rabbit anti-Human, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | ETHE1 |
---|---|
Clone | 1 |
Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Monoclonal |
Description
ETHE1 Monoclonal specifically detects ETHE1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
ETHE1 | |
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
23474 | |
IgG | |
Protein A purified |
1 | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
D83198, EC 3.1.2.6, ethylmalonic encephalopathy 1, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, HSCOEC 3.-, protein ETHE1, mitochondrial, YF13H12 | |
Recombinant Human ETHE1 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title