Learn More
Invitrogen™ ETHE1 Recombinant Rabbit Monoclonal Antibody (001)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529251
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human ETHE1/HSCO.
ETHE1 is a sulfur dioxygenase that localizes within the mitochondrial matrix. The enzyme functions in sulfide catabolism. Mutations in its gene result in ethylmalonic encephalopathy.
Specifications
ETHE1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O95571 | |
Ethe1 | |
Recombinant Human ETHE1/HSCO Protein (Leu13-Ala254). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
001 | |
Unconjugated | |
Ethe1 | |
0610025L15Rik; ETHE1; ETHE1, persulfide dioxygenase; ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; hepatoma subtracted clone one protein; HSCO; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; sulfur dioxygenase ETHE1; YF13H12 | |
Rabbit | |
Protein A | |
RUO | |
23474 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.