Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ethenoadenosine Monoclonal Antibody (1G4)
GREENER_CHOICE

Catalog No. 01676387
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-387 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-387 Supplier Invitrogen™ Supplier No. MA140268
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-40268 detects Ethenoadenosine. MA1-40268 has been successfully used in ELISA, flow cytometry, immunohistochemistry (frozen), immunoprecipitation, Western blot applications. The MA1-40268 immunogen is human ethenoadenosine.

The reaction of chloroacetaldehyde (CAA) with DNA results in the formation of ethenoadenosine also known as 1,N6-ethenodeoxy-adenosine or etheno-A. CAA is formed through metabolization of for example vinyl chloride, a well established carcinogen. In RNA, after in vitro activation, etheno-A and etheno-C are the principle products of RNA damage. Etheno derivate formation may be highest in single-stranded DNA-regions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ethenoadenosine
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1G4
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Alias C12H13N5O4; N6-Ethenoadenosine
Host Species Mouse
Immunogen Human ethenoadenosine.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.