Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethenocytidine Antibody (6F5), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600444B
Description
Ethenocytidine Monoclonal specifically detects Ethenocytidine in All Species samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Ethenocytidine | |
Monoclonal | |
Biotin | |
C11H13N3O5 | |
Ethenocytidine | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Cancer, Cell Biology | |
This is specific for ethenocytidine and ethenodecytidine. | |
Store at 4C in the dark. | |
IgG2 λ |
ELISA, Immunocytochemistry, Immunofluorescence | |
6F5 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction