Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethenoadenosine Antibody (1G4) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-4983929 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600442SS 0.025 mL
NB600442 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600442SS Supplier Novus Biologicals Supplier No. NB600442SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 16 publications

Ethenoadenosine Monoclonal specifically detects Ethenoadenosine in All Species samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.

Specifications

Antigen Ethenoadenosine
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF
Classification Monoclonal
Clone 1G4
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:800, Flow Cytometry 1:10-1:1000. Use reported in scientific literature (PMID 9774627), ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500, CyTOF-ready
Gene Alias C12H13N5O4, N6-Ethenoadenosine
Host Species Mouse
Immunogen Ethenoadenosine
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cell Biology
Primary or Secondary Primary
Test Specificity This is specific for ethenoadenosine and ethenodadenosine.
Target Species All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.