Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ethenocytidine Monoclonal Antibody (6F5)
GREENER_CHOICE

Catalog No. AMPA6886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPA6886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPA6886 Supplier Invitrogen™ Supplier No. MA116886
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: This is specific for ethenocytidine and ethenodecytidine.

Ethenocytidine, also known as epsilon cytidine, is a modified nucleoside derivative of cytidine, where an etheno bridge forms between the nitrogenous base and the sugar moiety. This structural alteration is best characterized through various spectroscopic methods such as NMR, which reveal distinct differences between its crystalline and solution states. Ethenocytidine is not directly encoded by genes but can be formed post-transcriptionally as a result of exposure to certain mutagenic chemicals. The presence of ethenocytidine can induce mutagenic effects due to its propensity to mispair during DNA replication, which can alter genetic coding and affect gene expression. Understanding these modifications is crucial in the study of carcinogenesis and the design of therapeutic interventions targeting gene transcription processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ethenocytidine
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Alias C11H13N3O5
Host Species Mouse
Immunogen Ethenocytidine.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2 λ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.