Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Anhydrous, Contains BHT as a Preservative, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5391126 Shop All Avantor Products
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Aluminum Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-817 250 mL Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-817 Supplier Avantor Supplier No. 924401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Flash Point −45.00°C
Packaging Aluminum Bottle
Quantity 250 mL
Specific Gravity 1L = 0.71kg
Formula Weight 74.12
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Ether, Anhydrous, Contains BHT as a Preservative
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.