Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Purified, Spectrum™ Chemical
SDP

Catalog No. 18610389 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Glass Bottle
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-389 20 L Glass Bottle
18-610-391 4 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-610-389 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E101620LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60-29-7 , 128-37-0
Molecular Weight (g/mol) 74.12

Specifications

CAS Max % 100%
Quantity 20 L
Packaging Glass Bottle
Boiling Point 34.6°C
Grade Purified
Chemical Name or Material Ether

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.