Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethidium bromide, >98%, Molecular Biology Reagent, MP Biomedicals™

Catalog No. ICN19399305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19399305 5 g
ICN19399301 1 g
ICN19399325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19399305 Supplier MP Biomedicals, Inc Supplier No. 0219399305
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1239-45-8
Molecular Formula C21H20BrN3
Molecular Weight (g/mol) 394.32
MDL Number MFCD00011724
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
Synonym ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
PubChem CID 14710
ChEBI CHEBI:4883
IUPAC Name 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Color Brown-Red
Assay Percent Range ≥98%
Quantity 5 g
Formula Weight 394.3
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material Ethidium bromide
EINECSNumber 214-984-6
Recommended Storage Store at room temperature (15 to 30°C).
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.