Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ethidium Bromide Solution (10mg/mL)

Catalog No. mp1etbc1001 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 mL
6 x 10 mL

Ethidium Bromide Solution (10mg/mL), Molecular biology grade

  • Concentration : 0.95 – 1.05%
  • Purity (on dry basis) : > 98.0%
  • RNase, DNase, Protease free
  • Fluorescent DNA and RNA intercalating agent.
  • Used as a dye for DNA and RNA electrophoresis.
  • Storage: Store at + 4°C protected from light.
  • A 10 mg/mL Ready-To-Use solution of EtBr in specially filtered, deionized water.
  • Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.
  • It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded D
  • Ethidium bromide (EtBr) is useful for all applications requiring nucleic acid staining including CsCl plasmid DNA isolation and is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1239-45-8
Detection 15-30 min with 0.5mg/mL solution
Color Dark Red/Purple
Concentration 10 mg/mL
Product Type Fluorescent DNA/RNA Stain
Form Liquid
For Use With (Application) Nucleic Acid Electrophoresis; Blotting & Detection; Nucleic Acidgel Electrophoresis & Blotting
Molecular Weight (g/mol) 394.3
Quantity 10 mL
Storage Requirements Store at +4°C, protect from light.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.