Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethinylestradiol, 98%, Thermo Scientific Chemicals

Catalog No. ac461550010
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-63-6
Molecular Formula C20H24O2
Molecular Weight (g/mol) 296.41
MDL Number MFCD00003690
InChI Key BFPYWIDHMRZLRN-SLHNCBLASA-N
Synonym ethinyl estradiol, ethynylestradiol, ethynyl estradiol, ethinylestradiol, ethinyloestradiol, ginestrene, 17-ethinylestradiol, progynon c, ethinoral, eticyclin
PubChem CID 5991
ChEBI CHEBI:4903
IUPAC Name (8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Melting Point 180.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 296.41
Percent Purity 98%
Chemical Name or Material Ethinylestradiol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed. May cause cancer.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 200-342-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.