Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butylurea, 98%, Thermo Scientific™
Click to view available options
:
25g; Glass bottle
Specifications
Specifications
CAS | 1118-12-3 |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C5H12N2O |
Linear Formula | (CH3)3CNHCONH2 |
Quantity | 25g |
MDL Number | MFCD00025448 |
Synonym | n-tert-butylurea, urea, 1,1-dimethylethyl, urea, tert-butyl, 1,1-dimethylethyl urea, urea, n-1,1-dimethylethyl, 1-tert-butylurea, t-butylurea, tert-butyl urea, tert-butyl-urea, tert-butyl isourea |
Solubility Information | Solubility in water: slightly soluble in water |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. May cause respiratory irritation. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor |
EINECSNumber | 214-257-3 |
RTECSNumber | YS3680000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction