Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free
Click to view available options
Quantity:
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer, Lewis acid

Specifications
Specifications
Color | White |
Melting Point | 169.0°C |
Heavy Metals (as Pb) | 50ppm max. |
Assay Percent Range | 99.5% |
Quantity | 1 kg |
Physical Form | Powder |
Linear Formula | H3BO3 |
Fieser | 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70 |
Merck Index | 15, 1339 |
Solubility Information | Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol. |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS P Statement May damage fertility. May damage the unborn child. |
Precautionary Statement | GHS P Statement: Immediately call a POISON CENTER or doctor/physician. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction