Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethoxymethylenemalononitrile, 98%

Catalog No. p-3778527
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle

CAS: 123-06-8 | C6H6N2O | 122.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-06-8
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00001854
InChI Key OEICGMPRFOJHKO-UHFFFAOYSA-N
Synonym ethoxymethylenemalononitrile, ethoxymethylene malononitrile, propanedinitrile, ethoxymethylene, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, 2-cyano-3-ethoxyacrylonitrile, ethoxymethylene propanedinitrile, usaf a-9230, usaf kf-10
PubChem CID 67152
IUPAC Name 2-(ethoxymethylidene)propanedinitrile
SMILES CCOC=C(C#N)C#N

Specifications

Melting Point 63°C to 68°C
Color Colorless to Yellow
Density 155g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 98%
Linear Formula C2H5OCH=C(CN)2
Beilstein 03, I, 162
Formula Weight 122.13
Percent Purity ≥97.5% (GC)
Chemical Name or Material Ethoxymethylenemalononitrile
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.