Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-(2,4-difluorophenyl)-7-Cl-6-F-4-O-hydropyridino[2,3-b]pyridine-3-carboxylate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
100491-29-0
C17H10ClF3N2O3
382.723
MFCD01863285
JLSXYCZRMYCIMY-UHFFFAOYSA-N
ethyl 7-chloro-1-2,4-difluorophenyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate,ethyl 7-chloro-1-2,4-difluorophenyl-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylate,7-chloro-1-2,4-difluorophenyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid ethyl ester,7-chloro-1-2,4-difluoro-phenyl-6-fluoro-4-oxo-1,4-dihydro-1,8 naphthyridine-3-carboxylic acid ethyl ester,ethyl 1-2,4-difluorophenyl-7-cl-6-f-4-o-hydropyridino 2,3-b pyridine-3-carboxylate,7-chloro-6-fluoro-1-2,4-difluorophenyl-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid ethyl ester,ethyl 1-2,4-difluorophenyl-7-chloro-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylate,ethyl 1-2,4-difluorophenyl-7-chloro-6-fluoro-4-oxohydropyridino 2,3-b pyridi ne-3-carboxylate,ethyl 1-2,4-difluorophenyl-7-chloro-6-fluoro-4-oxopyridino 2,3-b pyridine-3-carboxylate,ethyl 7-chloro-1-2,4-difluorophenyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylate
1268243
ethyl 7-chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylate
CCOC(=O)C1=CN(C2=NC(=C(C=C2C1=O)F)Cl)C3=C(C=C(C=C3)F)F
100491-29-0
C17H10ClF3N2O3
382.723
MFCD01863285
JLSXYCZRMYCIMY-UHFFFAOYSA-N
ethyl 7-chloro-1-2,4-difluorophenyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate,ethyl 7-chloro-1-2,4-difluorophenyl-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylate,7-chloro-1-2,4-difluorophenyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid ethyl ester,7-chloro-1-2,4-difluoro-phenyl-6-fluoro-4-oxo-1,4-dihydro-1,8 naphthyridine-3-carboxylic acid ethyl ester,ethyl 1-2,4-difluorophenyl-7-cl-6-f-4-o-hydropyridino 2,3-b pyridine-3-carboxylate,7-chloro-6-fluoro-1-2,4-difluorophenyl-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid ethyl ester,ethyl 1-2,4-difluorophenyl-7-chloro-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylate,ethyl 1-2,4-difluorophenyl-7-chloro-6-fluoro-4-oxohydropyridino 2,3-b pyridi ne-3-carboxylate,ethyl 1-2,4-difluorophenyl-7-chloro-6-fluoro-4-oxopyridino 2,3-b pyridine-3-carboxylate,ethyl 7-chloro-1-2,4-difluorophenyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylate
1268243
ethyl 7-chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylate
CCOC(=O)C1=CN(C2=NC(=C(C=C2C1=O)F)Cl)C3=C(C=C(C=C3)F)F
Melting Point 212°C to 215°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color White
Assay Percent Range 99%
Formula Weight 382.73
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material Ethyl 1-(2, 4-difluorophenyl)-7-Cl-6-F-4-O-hydropyridino[2, 3-b]pyridine-3-carboxylate, 99%

GHS H Statement:
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning