Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl 1,3-dithiolane-2-carboxylate, 99%, ACROS Organics™
Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
Specifications
Boiling Point | 85°C (0.1mmHg) |
Specific Gravity | 1.24 |
Flash Point | 110°C |
Refractive Index | 1.5380 to 1.54 |
Density | 1.2400g/mL |
Packaging | Glass bottle |
Quantity | 5mL |
Color | Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Show More |
Safety and Handling
EINECSNumber : 243-837-9