Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylate 97.0+%, TCI America™
SDP

Catalog No. B274025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B274025G 25 g
B27405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B274025G Supplier TCI America Supplier No. B274025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142851-03-4
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD01763998
InChI Key MYHJCTUTPIKNAT-UHFFFAOYSA-N
Synonym ethyl n-boc-piperidine-4-carboxylate, 1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate, ethyl n-boc-4-piperidinecarboxylate, n-boc-4-carbethoxy piperidine, 1,4-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 4-ethyl ester, ethyl n-boc-isonipecotate, ethyl 1-boc-piperidine-4-carboxylate, dimethylethyl piperidinedicarboxylic acid ethyl ester, 1-boc-piperidine-4-carboxylic acid ethyl ester, 1-boc-isonipecotic acid ethyl ester
PubChem CID 2758812
IUPAC Name 1-O-tert-butyl 4-O-ethyl piperidine-1,4-dicarboxylate
SMILES CCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 g
Formula Weight 257.33
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.