Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Ethyl 2,3-butadienoate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g
5 g
Description
- Ethyl 2,3-butadienoate is an α-allenic ester
- The reaction of ethyl 2,3-butadienoate with n-tosylated imines in the presence of dabco (1,4-diazabicyclo[2.2.2]octane) or dmap (4-dimethylaminopyridine) forms azetidine derivatives or novel dihydropyridine derivatives respectively
- The performance of bifunctional n-acyl aminophosphines to catalyze the asymmetric [3+2] cycloaddition of phenylidenemalononitrile with ethyl 2,3-butadienoate has been evaluated.ethyl 2,3-butadienoate may be used in the synthesis of dihydropyrans by reacting with acyclic enones
- It may also be used to synthesize spiranic heterocycles by reacting with heterocyclic bis-arylidene ketones via phosphine-catalyzed [3+2] annulations
Specifications
Specifications
| CAS | 14369-81-4 |
| Density | 0.966 g/mL (at 25°C (literature)) |
| Boiling Point | 80°C (60 mmHg) |
| Molecular Formula | C6H8O2 |
| Refractive Index | n20/D 1.46 (literature) |
| Linear Formula | H2C=C=CHCO2C2H5 |
| MDL Number | MFCD01863567 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 112.13 |
| Percent Purity | 95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction