Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-4-(2-furyl)thiophene-3-carboxylate, 97%, Thermo Scientific™

Catalog No. p-7039478
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5873304 2 g
AAH5873309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5873304 Supplier Thermo Scientific Chemicals Supplier No. H5873304

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256506-99-7
Molecular Formula C11H11NO3S
Molecular Weight (g/mol) 237.27
MDL Number MFCD00126394
InChI Key JMLBJOYPNPOBKV-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-2-furyl thiophene-3-carboxylate, ethyl 2-amino-4-furan-2-yl thiophene-3-carboxylate, 3-thiophenecarboxylicacid, 2-amino-4-2-furanyl-, ethyl ester, ethyl 2-amino-4-2-furyl-3-thiophenecarboxylate, ethyl 2-azanyl-4-furan-2-yl thiophene-3-carboxylate, 2-amino-4-2-furanyl-3-thiophenecarboxylic acid ethyl ester, 2-amino-4-furan-2-yl-thiophene-3-carboxylic acid ethyl ester
PubChem CID 736628
IUPAC Name ethyl 2-amino-4-(furan-2-yl)thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(N)SC=C1C1=CC=CO1

Specifications

Quantity 2 g
Formula Weight 237.27
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-(2-furyl)thiophene-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.