Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-4-(4-acetoxyphenyl)thiophene-3-carboxylate, 97%, Thermo Scientific™

Catalog No. AAH5874204
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5874204 2 g
AAH5874209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5874204 Supplier Thermo Scientific Chemicals Supplier No. H5874204

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858250-46-0
Molecular Formula C15H15NO4S
Molecular Weight (g/mol) 305.35
MDL Number MFCD22988973
InChI Key RAXUJVOBTPWILV-UHFFFAOYSA-N
Synonym ethyl 4-4-acetyloxy phenyl-2-aminothiophene-3-carboxylate, ethyl 2-amino-4-4-acetoxyphenyl thiophene-3-carboxylate
PubChem CID 98000080
SMILES CCOC(=O)C1=C(N)SC=C1C1=CC=C(OC(C)=O)C=C1

Specifications

Quantity 2 g
IUPAC Name ethyl 4-(4-acetyloxyphenyl)-2-aminothiophene-3-carboxylate
Formula Weight 305.34
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-(4-acetoxyphenyl)thiophene-3-carboxylate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.