Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-4-phenyl-5-thiazolecarboxylate, 99%, Thermo Scientific™

Catalog No. ac162220025
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 64399-23-1
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD00022452
InChI Key OZMXFXOHCUEEPD-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-phenylthiazole-5-carboxylate, ethyl 2-amino-4-phenyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-amino-4-phenyl-, ethyl ester, 2-amino-4-phenylthiazole-5-carboxylic acid ethyl ester, acmc-209nlv, 2-amino-4-phenyl-5-carboethoxythiazole, ethyl 2-amino-4-phenylthiazol-5-carboxylate, ethyl2-amino-4-phenylthiazole-5-carboxylate, ethyl 2-amino-4-phenyl-thiazole-5-carboxylate, ethyl 2-amino-4-phenylthiazol-5-yl-carboxylate
PubChem CID 73239
IUPAC Name ethyl 2-amino-4-phenyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)N)C2=CC=CC=C2

Specifications

Melting Point 169°C to 172°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 2.5 g
Formula Weight 248.3
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-amino-4-phenyl-5-thiazolecarboxylate, 99%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.