Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-4-thiazoleacetate, 99%, Thermo Scientific™

Catalog No. AC205300500
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass Bottle
Plastic bottle

Chemical Identifiers

CAS 53266-94-7
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00005330
InChI Key SHQNGLYXRFCPGZ-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate
PubChem CID 104454
IUPAC Name ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
SMILES CCOC(=O)CC1=CSC(N)=N1

Specifications

Melting Point 91.0°C to 95.0°C
Color Yellow to Beige
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 50 g
Formula Weight 186.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-amino-4-thiazoleacetate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.