Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Bromovalerate 98.0+%, TCI America™
SDP

Catalog No. E0970100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0970100G 100 g
E097025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0970100G Supplier TCI America Supplier No. E0970100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 615-83-8
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00000159
InChI Key ORSIRXYHFPHWTN-UHFFFAOYSA-N
PubChem CID 79071
IUPAC Name ethyl 2-bromopentanoate
SMILES CCCC(C(=O)OCC)Br

Specifications

Color Yellow
Boiling Point 94°C
Quantity 100 g
UN Number 3265
Synonym ethyl 2-bromovalerate, ethyl2-bromovalerate, ethyl .alpha.-bromovalerate, pentanoic acid, 2-bromo-, ethyl ester, 2-bromovaleric acid ethyl ester, 2-bromopentanoic acid ethyl ester, 2-bromo-pentanoic acid ethyl ester, pentanoic acid, bromo-, ethyl ester, ethyl bromovalerate, ethyl-2bromovalerate
Formula Weight 209.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Bromovalerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.