Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Butylacetoacetate 96.0+%, TCI America™
SDP

Catalog No. B070225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B070225ML 25 mL
B07025ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B070225ML Supplier TCI America Supplier No. B070225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1540-29-0
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.251
MDL Number MFCD00026888
InChI Key ZTOQBHVLCJERBS-UHFFFAOYSA-N
Synonym ethyl 2-n-butylacetoacetate, ethyl 2-butylacetoacetate, hexanoic acid, 2-acetyl-, ethyl ester, ethyl .alpha.-butylacetoacetate, 2-acetylhexanoic acid ethyl ester, ethyl butyl acetoacetate, ethyl 2-acetyl hexanoate, acmc-209da5, ethyl-2-n-butylacetoacetate, ksc494i1n
PubChem CID 95466
IUPAC Name ethyl 2-acetylhexanoate
SMILES CCCCC(C(=O)C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 224°C
Quantity 25 mL
Formula Weight 186.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Butylacetoacetate
EINECSNumber (2)-1505
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.