Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-butynoate

Catalog No. 4251171G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g
10 g

Catalog No. 42-511-71G

Supplier: Sigma Aldrich 4251171G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 2-butynoate, a 2-alkynoate is a methanogenesis inhibitor. it is an electron deficient internal alkyne that has been reported to undergo codimerization with alkenes to form 1,3-dienes catalyzed by rhodium(i)/h8-binap complex. the annulation of thioamides with ethyl 2-butynoate catalyzed by tri-n-butylphosphine to form thiazolines has been investigated.ethyl 2-butynoate was used in the study of its effect on in vitro degradation and microbial biomass production.it may be used in the synthesis of the following:
  • 3-ethyl, 1-methyl 1-(2-nitrophenyl)-cyclopent-3-ene-1,3-dicarboxylate
  • Tricyclic aziridine derivatives
  • Alkenylsilanols

Specifications

CAS 4341-76-8
Density 0.962 g/mL (at 25°C (literature))
Boiling Point 160°C to 161°C (730 mmHg)
Molecular Formula C6H8O2
Refractive Index n20/D 1.436 (literature)
Linear Formula CH3C///CC(O)OCH2CH3
MDL Number MFCD00015182
Quantity 1 g
Synonym Ethyl tetrolate
Molecular Weight (g/mol) 112.13
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.