Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-cyano-2-methylpropionate, 97%

Catalog No. p-7030668
Click to view available options
Quantity:
10 g
50 g

CAS: 1572-98-1 | C7H11NO2 | 141.17 g/mol

It is employed as a intermediates for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical and organic synthesis.

Solubility
Soluble in ether. Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1572-98-1
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00034667
InChI Key FYGRPGOHQCPZCV-UHFFFAOYSA-N
PubChem CID 344887
IUPAC Name ethyl 2-cyano-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)C#N

Specifications

Density 0.97
Boiling Point 185°C
Quantity 10 g
Synonym 2-cyano-2-methylpropionic acid ethyl ester, ethyl 2-cyano-2-methylpropionate, ethyl 2-cyano-2,2-dimethylacetate, 2-cyano-2-methylpropanoic acid ethyl ester, ethyl 2-cyano-2-methyl-propanoate, propanoic acid, 2-cyano-2-methyl-, ethyl ester, pubchem23352, acmc-209dfv, ksc529g3h, ethyl2-cyano-2-methylpropionate
Solubility Information Soluble in ether. Not miscible or difficult to mix with water.
Formula Weight 141.17
Percent Purity 97%
Chemical Name or Material Ethyl 2-cyano-2-methylpropionate
Hazard Category H302-H318
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.