Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-cyanoacetoacetate

Catalog No. AA4379204
Click to view available options
Quantity:
2 g
10 g

Catalog No. AA4379204

Supplier: Thermo Scientific Chemicals 04379204

Only null left
Add to Cart

CAS: 634-55-9 | C7H9NO3 | 155.15 g/mol

It is used as a pharmaceutical intermediate. It is also used in the synthesis of fluorine-containing alkyl 2-cyano-3-oxocarboxylates and also in the syntheses of pyrazolo [1, 2-a] pyrazole and pyrazolo [5, 1-b] [1, 3] oxazine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-55-9
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.15
MDL Number MFCD00043487
InChI Key NWOKVFOTWMZMHL-UHFFFAOYNA-N
Synonym ethyl 2-cyanoacetoacetate, ethyl 2-acetylcyanoacetate, ethyl a-cyanoacetoacetate, ethyl alpha-cyanoacetoacetate, butanoic acid, 2-cyano-3-oxo-, ethyl ester, ethyl2-acetylcyanoacetate, ethyl 2-cyano-3-oxo-butanoate, 2-cyanoacetoacetic acid ethyl ester, 2-cyano-3-oxo-butyric acid ethyl ester, zlchem 939
PubChem CID 136455
ChEBI CHEBI:51924
IUPAC Name ethyl 2-cyano-3-oxobutanoate
SMILES CCOC(=O)C(C#N)C(C)=O

Specifications

Quantity 2 g
UN Number UN3276
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 155.16
Physical Form Liquid
Chemical Name or Material Ethyl 2-cyanoacetoacetate
Hazard Category H301+H311-H330
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.