Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-cyclohexanoneacetate, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
24731-17-7
C10H16O3
184.23
MFCD00001638
ZZWSNYNCRUZSPR-UHFFFAOYSA-N
ethyl 2-2-oxocyclohexyl acetate,ethyl 2-cyclohexanoneacetate,ethyl 2-oxocyclohexaneacetate,ethyl 2-oxocyclohexyl acetate,ethyl 2-cyclohexanone acetate,ethyl-2-cyclohexanone acetate,ethyl 2-oxocyclohexylacetate,cyclohexaneacetic acid, 2-oxo-, ethyl ester,ethyl 2-oxo-cyclohexyl-acetate
91229
ethyl 2-(2-oxocyclohexyl)acetate
CCOC(=O)CC1CCCCC1=O
24731-17-7
C10H16O3
184.23
MFCD00001638
ZZWSNYNCRUZSPR-UHFFFAOYSA-N
ethyl 2-2-oxocyclohexyl acetate,ethyl 2-cyclohexanoneacetate,ethyl 2-oxocyclohexaneacetate,ethyl 2-oxocyclohexyl acetate,ethyl 2-cyclohexanone acetate,ethyl-2-cyclohexanone acetate,ethyl 2-oxocyclohexylacetate,cyclohexaneacetic acid, 2-oxo-, ethyl ester,ethyl 2-oxo-cyclohexyl-acetate
91229
ethyl 2-(2-oxocyclohexyl)acetate
CCOC(=O)CC1CCCCC1=O
Flash Point >110°C
Specific Gravity 1.02
Infrared Spectrum Authentic
Density 1.0200g/mL
Color Colorless to Yellow
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Quantity 5g
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula (O=)C6H9CH2CO2C2H5
Formula Weight 184.23
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Ethyl 2-cyclohexanoneacetate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.