Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-hydroxyethyl sulfide, 98%

Catalog No. p-130120
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

CAS: 110-77-0 | C4H10OS | 106.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-77-0
Molecular Formula C4H10OS
Molecular Weight (g/mol) 106.18
MDL Number MFCD00002909
InChI Key LNRIEBFNWGMXKP-UHFFFAOYSA-N
Synonym 2-ethylthio ethanol, ethyl 2-hydroxyethyl sulfide, ethanol, 2-ethylthio, ethyl thioethanol, 2-hydroxyethyl ethyl sulfide, ethyl 2-hydroxyethyl thioether, beta-ethylthioethanol, 2-ethylsulfanyl ethanol, beta-hydroxydiethyl sulfide, 2-ethylthioethanol
PubChem CID 8075
IUPAC Name 2-ethylsulfanylethanol
SMILES CCSCCO

Specifications

Melting Point -100°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 180°C to 184°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4859 to 1.4879
Linear Formula C2H5SCH2CH2OH
Quantity 5 g
Beilstein 01, II, 525
Specific Gravity 1.02
Merck Index 15, 3906
Formula Weight 106.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 2-hydroxyethyl sulfide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
EINECSNumber 203-802-0
RTECSNumber KL1225000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.