Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-methylacetoacetate, 95%

Catalog No. p-130789
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 609-14-3 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-14-3
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009164
InChI Key FNENWZWNOPCZGK-UHFFFAOYSA-N
Synonym ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
PubChem CID 701
IUPAC Name ethyl 2-methyl-3-oxobutanoate
SMILES CCOC(=O)C(C)C(=O)C

Specifications

Melting Point -45°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 187°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4170 to 1.419
Linear Formula CH3COCH(CH3)CO2C2H5
Quantity 25 g
Beilstein 03, 679
Specific Gravity 1.01
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents
Formula Weight 144.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 2-methylacetoacetate, 95%
Show More Show Less
EINECSNumber 210-179-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.