Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-(N-phthalimidooxy)-2-methylpropionate, 98%, Thermo Scientific™

Catalog No. AAH6166003
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40674-21-3
Molecular Formula C14H15NO5
Molecular Weight (g/mol) 277.276
InChI Key UNZUMKCIIIFEJI-UHFFFAOYSA-N
Synonym ethyl 2-1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl oxy-2-methylpropionate, ethyl 2-1,3-dioxoisoindolin-2-yloxy-2-methylpropanoate, ethyl 2-1,3-dioxoisoindol-2-yl oxy-2-methylpropanoate, 2-phthalimidyloxy isobutyric acid ethyl ester, propanoic acid,2-1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl oxy-2-methyl-, ethyl ester
PubChem CID 3016209
IUPAC Name ethyl 2-(1,3-dioxoisoindol-2-yl)oxy-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)ON1C(=O)C2=CC=CC=C2C1=O

Specifications

Quantity 1 g
Formula Weight 277.27
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl 2-(N-phthalimidooxy)-2-methylpropionate
EINECSNumber 255-030-9
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.