Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-nitrobenzoate, 97%

Catalog No. p-7024781
Click to view available options
Quantity:
25 g
100 g

CAS: 610-34-4 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-34-4
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00014696
InChI Key CPNMAYYYYSWTIV-UHFFFAOYSA-N
Synonym ethyl o-nitrobenzoate, ethyl nitrobenzoate, benzoic acid, o-nitro-, ethyl ester, benzoic acid, 2-nitro-, ethyl ester, nitrobenzoesaureathylester, acmc-1aul6, 2-nitro-benzoic acid ethyl ester
PubChem CID 69123
IUPAC Name ethyl 2-nitrobenzoate
SMILES CCOC(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 26°C to 30°C
Boiling Point 172°C to 174°C (18 mmHg)
Quantity 25 g
Beilstein 2050729
Merck Index 14,3832
Formula Weight 195.18
Percent Purity 97%
Chemical Name or Material Ethyl 2-nitrobenzoate
EINECSNumber 210-220-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.