Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Oxocyclopentanecarboxylate 97.0+%, TCI America™
SDP

Catalog No. B0268250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0268250ML 250 mL
B026825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0268250ML Supplier TCI America Supplier No. B0268250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 611-10-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00001412
InChI Key JHZPNBKZPAWCJD-UHFFFAOYSA-N
Synonym ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate
PubChem CID 69136
IUPAC Name ethyl 2-oxocyclopentane-1-carboxylate
SMILES CCOC(=O)C1CCCC1=O

Specifications

Color Yellow
Boiling Point 132°C
Quantity 250 mL
Formula Weight 156.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Oxocyclopentanecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.