Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-thiopheneacetate, 98%, Thermo Scientific™

Catalog No. AC303770250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57382-97-5
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.23
MDL Number MFCD00005457
InChI Key QSUANHXENVRFDN-UHFFFAOYSA-N
Synonym ethyl 2-thiopheneacetate, ethyl 2-thienylacetate, ethyl 2-thiophen-2-yl acetate, ethyl thiophene-2-acetate, ethyl thiophen-2-ylacetate, ethyl 2-2-thienyl acetate, ethyl-2-thiophene acetate, thiopheneacetic acid, ethyl ester, 2-thiopheneacetic acid, ethyl ester, pubchem7414
PubChem CID 93657
IUPAC Name ethyl 2-(thiophen-2-yl)acetate
SMILES CCOC(=O)CC1=CC=CS1

Specifications

Color Colorless to Yellow
Density 1.1300g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5090 to 1.5110 (20°C, 589nm)
Quantity 25 g
Formula Weight 170.23
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Ethyl 2-thiopheneacetate, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.