Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate, Thermo Scientific™

Catalog No. MAY00183DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00183DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00183DA Supplier Thermo Scientific Chemicals Supplier No. MAY00183DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 306960-67-8
Molecular Formula C9H9F3N2O2
Molecular Weight (g/mol) 234.178
InChI Key LYEKBQSEKSEXLT-UHFFFAOYSA-N
Synonym ethyl 4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, ethyl 2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, ethyl-2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-methyl-2-trifluoromethyl-, ethyl ester
PubChem CID 10609709
IUPAC Name ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1C)C(F)(F)F

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 234.18
Percent Purity 97%
Chemical Name or Material Ethyl 2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.