Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3,3-Diethoxyacrylate 95.0+%, TCI America™
SDP

Catalog No. D200625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D200625G 25 g
D20065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D200625G Supplier TCI America Supplier No. D200625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32002-24-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00012220
InChI Key POJGRKZMYVJCST-UHFFFAOYSA-N
Synonym 3,3-Diethoxyacrylic Acid Ethyl Ester
PubChem CID 169388
IUPAC Name ethyl 3,3-diethoxyprop-2-enoate
SMILES CCOC(=CC(=O)OCC)OCC

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 g
Formula Weight 188.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3,3-Diethoxyacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.