Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3,5-Dimethyl-2-pyrrolecarboxylate 98.0+%, TCI America™
SDP

Catalog No. E084325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E084325G 25 g
E08435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E084325G Supplier TCI America Supplier No. E084325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2199-44-2
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00010645
InChI Key IZSBSZYFPYIJDI-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, ethyl 3,5-dimethylpyrrole-2-carboxylate, ethyl 3,5-dimethyl-2-pyrrolecarboxylate, 1h-pyrrole-2-carboxylic acid, 3,5-dimethyl-, ethyl ester, 3,5-dimethyl-1h-pyrrole-2-carboxylic acidethyl ester, ethyl 3,5-dimethyl-1h-pyrrole-2-carboxylate, pubchem23984, acmc-1cdc7, 3,5-dimethyl-1h-pyrrole-2-carboxylicacidethylester, ksc496q7t
PubChem CID 137477
IUPAC Name ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=C(C=C(N1)C)C

Specifications

Melting Point 124°C
Boiling Point 272°C
Quantity 25 g
Formula Weight 167.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3,5-Dimethyl-2-pyrrolecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.