Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-bromobenzoate, 98+%

Catalog No. p-7080637
Click to view available options
Quantity:
10 g
50 g

CAS: 24398-88-7 | C9H9BrO2 | 229.073 g/mol

Ethyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4'-methyl-biphenyl-3-carboxylic acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4′-methyl-biphenyl-3-carboxylic acid ethyl ester.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24398-88-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00013529
InChI Key QAUASTLEZAPQTB-UHFFFAOYSA-N
PubChem CID 90488
IUPAC Name ethyl 3-bromobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)Br

Specifications

Density 1.43
Boiling Point 261°C
Flash Point >110°C (230°F)
Refractive Index 1.543
Quantity 10 g
Beilstein 1867121
Synonym ethyl3-bromobenzoate, 3-bromobenzoic acid ethyl ester, benzoic acid, 3-bromo-, ethyl ester, ethyl-3-bromobenzoate, benzoic acid, m-bromo-, ethyl ester, m-bromo benzoic acid ethyl ester, m-bromobenzoic acid, ethyl ester, pubchem3941, acmc-209gbv, ethyl 3-bromobenzoate
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-bromobenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-223-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.