Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Cyclohexene-1-carboxylate 95.0+%, TCI America™
SDP

Catalog No. C096625ML
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 15111-56-5
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00037158
InChI Key BYNLHLLOIZDKIC-UHFFFAOYSA-N
Synonym 3-Cyclohexene-1-carboxylic Acid Ethyl Ester
PubChem CID 85803
IUPAC Name ethyl cyclohex-3-ene-1-carboxylate
SMILES CCOC(=O)C1CCC=CC1

Specifications

Color Yellow
Boiling Point 195°C
Quantity 25 mL
Formula Weight 154.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Cyclohexene-1-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.