Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-ethoxyacrylate, 98%

Catalog No. p-7053149
Click to view available options
Quantity:
10 g
50 g

CAS: 1001-26-9 | C7H12O3 | 144.17 g/mol

Used in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1001-26-9
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009863
InChI Key ITQFPVUDTFABDH-AATRIKPKSA-N
Synonym ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate
PubChem CID 5366269
IUPAC Name ethyl (E)-3-ethoxyprop-2-enoate
SMILES CCOC=CC(=O)OCC

Specifications

Color Colorless
Density 0.998 g/mL
Boiling Point 195°C to 196°C
Flash Point 76°C (169°F)
Linear Formula CH3CH2OCH=CHCOOCH2CH3
Refractive Index 1.4465
Quantity 10 g
UN Number UN3334
Beilstein 1721612
Solubility Information Slightly soluble in water.
Formula Weight 144.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3-ethoxyacrylate
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.