Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Ethoxyacrylate (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. E004125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E004125ML 25 mL
E00415ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E004125ML Supplier TCI America Supplier No. E004125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1001-26-9
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009863
InChI Key ITQFPVUDTFABDH-AATRIKPKSA-N
PubChem CID 5366269
IUPAC Name ethyl (E)-3-ethoxyprop-2-enoate
SMILES CCOC=CC(=O)OCC

Specifications

Color Yellow
Quantity 25 mL
Synonym ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate
Formula Weight 144.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Ethoxyacrylate (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.